Order Entry
Germany
ContactUsLinkComponent
DL-Ornithine hydrochloride, Sigma-Aldrich®
DL-Ornithine hydrochloride, Sigma-Aldrich®
Catalog # SIAL75490-10G
Supplier:  Merck
CAS Number:  
DL-Ornithine hydrochloride, Sigma-Aldrich®
Catalog # SIAL75490-10G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
 :168.62 g/mol
EINECS:213-956-0
CAS Number:1069-31-4
Formula:C₅H₁₂N₂O₂·HCl
MDL Number:MFCD00065398