DL-Ornithine hydrochloride ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1069-31-4
:
Formula:C₅H₁₂N₂O₂·HCl
MDL Number:MFCD00065398
:168.62 g/mol
Melting Pt:∼ 225 °C (dec.)
Storage Temperature:Ambient
EINECS:213-956-0