Order Entry
Germany
ContactUsLinkComponent
 
Arsenic standard solution, 1000 mg/L As in 2% hydrochloric acid (from As₂O₃) Standard for ICP, Assurance Grade
Catalog # JOBISPEC-AS3
CAS Number:  
Arsenic standard solution, 1000 mg/L As in 2% hydrochloric acid (from As₂O₃) Standard for ICP, Assurance Grade
Catalog # JOBISPEC-AS3
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    2% HCl
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Description:
    ICP standard As III
  • Element:
    Arsenic
  • Symbol:
    As

Specifications

About this item

Certifications: Traceable to SRM from NIST, tested in ISO 17034 / ISO 17025 accredited laboratory.