Antimony standard solution, 1000 mg/L Sb in dil. nitric acid with hydrofluoric acid (max. 1%) standard for AAS and ICP
Catalog # CPIIS4400-100023
Supplier: CPI
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:2-5% HNO₃+tr. HF
- Pack type:Glass bottle
- Pk:100 mL
- Element:Antimony
- Symbol:Sb