Order Entry
Germany
ContactUsLinkComponent
Antimony standard solution, 1000 mg/L Sb in 1% nitric acid with 0,6% tartaric acid (from Sb) standard for ICP-MS
Antimony standard solution, 1000 mg/L Sb in 1% nitric acid with 0,6% tartaric acid (from Sb) standard for ICP-MS
Catalog # JOBIJYICP-SB
CAS Number:  
Antimony standard solution, 1000 mg/L Sb in 1% nitric acid with 0,6% tartaric acid (from Sb) standard for ICP-MS
Catalog # JOBIJYICP-SB
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    H₂O/tr HNO₃/tr tartaric acid
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Element:
    Antimony
  • Symbol:
    Sb

Specifications

About this item

Certifications: Traceable to SRM from NIST, tested in ISO 17034 / ISO 17025 accredited laboratory.