Order Entry
Germany
ContactUsLinkComponent
Sulphuric acid ≥95.50% calibration solution of viscosity acc. to DIN EN ISO 307
Corrosive
Sulphuric acid ≥95.50% calibration solution of viscosity acc. to DIN EN ISO 307
Catalog # KRAF16350.3700
Supplier:  Bernd Kraft
CAS Number:  
Corrosive
Sulphuric acid ≥95.50% calibration solution of viscosity acc. to DIN EN ISO 307
Catalog # KRAF16350.3700
Supplier:  Bernd Kraft
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :98.08 g/mol
Merck Index:14,08974
Boiling Pt:∼335 °C (1013 hPa)
Melting Pt:3 °C
Density:1.84 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-639-5
UN:1830
ADR:8,II
 :GHS05
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589