Order Entry
Germany
Orders LinkContactUsLinkComponent
Ammonia solution 15%, extra pure

Ammonia solution 15%, extra pure

Corrosive
Irritant
Environment
Supplier:   Bernd Kraft
Ammonia solution 15%, extra pure
Supplier:  Bernd Kraft
Certificates
Ammonia solution 15%, extra pure

3 Options Available Below

Product Details & Documents
Synonyms:  Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
  • CAS Number:1336-21-6
  • Formula:NH₃
  • MDL Number:MFCD00066650
  • Molecular Weight:35.05 g/mol
  • Density:0.92 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-647-6
  • UN:2672
  • ADR:8,III
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Plastic bottle
    2.500 mL
    Log in for availability
    Log in for availability
    1 * 2.500 mL
    1 * 2.500 mL - 126,00 €
    126,00 €
    1 * 2.500 mL
       126,00 €

  • Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 92,00 €
    92,00 €
    1 * 1 L
       92,00 €

  • Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 170,00 €
    170,00 €
    1 * 5 L
       170,00 €