Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Sulphuric acid
Sulphuric acid 62.3%, extra pure for the determination of fats in milk and cheese
Sulphuric acid 62.3%, extra pure for the determination of fats in milk and cheese
:
Bernd Kraft
Sulphuric acid 62.3%, extra pure for the determination of fats in milk and cheese
:
Bernd Kraft
:
7664-93-9
:
:
H₂SO₄
:
MFCD00064589
:
98.08 g/mol
:
∼100 °C (1013 hPa)
:
1.5…1.8 g/cm³ (20 °C)
:
Ambient
:
231-639-5
:
1830
:
8,II
:
GHS05