Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Orthophosphoric acid
Orthophosphoric acid ≥85% analytical reagent
Orthophosphoric acid ≥85% analytical reagent
Orthophosphoric acid ≥85% analytical reagent
Supplier:
Bernd Kraft
Certificates
Expand image
Orthophosphoric acid ≥85% analytical reagent
Supplier:
Bernd Kraft
About this item
CAS Number:
7664-38-2
:
Formula:
H₃PO₄
MDL Number:
MFCD00011340
:
98 g/mol
Storage Temperature:
Ambient
EINECS:
231-633-2
UN:
1805
ADR:
8,III