Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Orthophosphoric acid
Orthophosphoric acid 35%
Orthophosphoric acid 35%
OVER838129
:
OVERLACK CHEMISCHE PRODUKTE
:
Orthophosphoric acid 35%
OVER838129
:
OVERLACK CHEMISCHE PRODUKTE
:
Pk:
25 kg
Seq:
4283
:
:
98 g/mol
:
1.5…1.8 g/cm³ (20 °C)
:
Ambient
:
231-633-2
:
1805
:
8,III
:
GHS05
:
7664-38-2
:
H₃PO₄
:
MFCD00011340