Sulphuric acid 0.3 mol/l (0.6 N) in aqueous solution for analysis of silicates
Catalog # KRAF06114.6020
Supplier: Bernd Kraft
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:10 L
- Seq:5214
Specifications
About this item
:
:98.08 g/mol
Density:1.01 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-639-5
UN:2796
ADR:8,II
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589