Order Entry
Germany
ContactUsLinkComponent
 
Aluminium standard solution, 10000 mg/L Al in 5% hydrochloric acid (from Al) standard for AAS and ICP
Catalog # JOBIPLAL1-3X
CAS Number:  
Aluminium standard solution, 10000 mg/L Al in 5% hydrochloric acid (from Al) standard for AAS and ICP
Catalog # JOBIPLAL1-3X
CAS Number:  

Specifications

  • Concentration:
    10000 mg/L
  • Matrix:
    5% HCl
  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Element:
    Aluminium
  • Symbol:
    Al

Specifications

About this item

Certifications: Traceable to SRM from NIST, tested in ISO 17034 / ISO 17025 accredited laboratory.