Lead standard solution, 5000 mg/L Pb in 1 M nitric acid (from Pb) standard for AAS
Catalog # KRAF03457.2000
Supplier: Bernd Kraft
CAS Number:
Specifications
- Concentration:5000 mg/L
- Matrix:1 M HNO₃
- Pack type:Plastic bottle
- Pk:500 mL
- Element:Lead
- Symbol:Pb
Specifications
About this item
Certifications: Traceable to NIST