Order Entry
Germany
ContactUsLinkComponent
Lead standard solution, 10000 mg/L Pb in 2 M nitric acid (from Pb) standard for ICP
Lead standard solution, 10000 mg/L Pb in 2 M nitric acid (from Pb) standard for ICP
Catalog # KRAF12904.0000
Supplier:  Bernd Kraft
CAS Number:  
Lead standard solution, 10000 mg/L Pb in 2 M nitric acid (from Pb) standard for ICP
Catalog # KRAF12904.0000
Supplier:  Bernd Kraft
CAS Number:  

Specifications

  • Concentration:
    10000 mg/L
  • Matrix:
    2 M HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Lead
  • Symbol:
    Pb

Specifications

About this item