Order Entry
Germany
ContactUsLinkComponent
 
Antimony standard solution, 1000 mg/L Sb in 5 M hydrochloric acid (from Sb) standard for ICP
  KRAF03851.0000
 :  
Antimony standard solution, 1000 mg/L Sb in 5 M hydrochloric acid (from Sb) standard for ICP
  KRAF03851.0000
 :  

 

  • Concentration:
    1000 mg/L
  • Matrix:
    5 M HCl
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Antimony
  • Symbol:
    Sb

 

 

 : Traceable to NIST.