Antimony standard solution, 10000 mg/L Sb in 5 M hydrochloric acid (from Sb₂O₃) standard for ICP
Catalog # KRAF12943.0000
Supplier: Bernd Kraft
CAS Number:
Specifications
- Concentration:10000 mg/L
- Matrix:5 M HCl
- Pack type:Plastic bottle
- Pk:100 mL
- Element:Antimony
- Symbol:Sb