Dipotassium oxalate 0.25 mol/l in aqueous solution
Supplier: Bernd Kraft
Certificates
About this item
CAS Number:583-52-8
:
Formula:C₂K₂O₄
MDL Number:MFCD00012464
:166.22 g/mol
Merck Index:12,07820
Storage Temperature:Ambient
EINECS:209-506-8
Frequently Bought Together