Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Pyridoxal hydrochloride
Pyridoxal hydrochloride, Sigma-Aldrich®
Pyridoxal hydrochloride, Sigma-Aldrich®
Pyridoxal hydrochloride, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
Pyridoxal hydrochloride, Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
65-22-5
:
Formula:
C₈H₁₀ClNO₃
MDL Number:
MFCD00012809
:
203.62 g/mol
EINECS:
200-602-5