Pyridoxal hydrochloride
Catalog # MOLE13958515-25G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:203.62 g/mol
EINECS:200-602-5
CAS Number:65-22-5
Formula:C₈H₁₀ClNO₃
MDL Number:MFCD00012809