
Sodium hydroxide 0.4 mol/l (0.4 N) in aqueous solution analytical reagent
Catalog # KRAF20653.3000
Supplier: Bernd Kraft
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 L
- Seq:5037
Specifications
About this item
:
:40 g/mol
Boiling Pt:∼105 °C (1013 hPa)
Density:1.02 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:1824
ADR:8,III
:GHS07
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548