Ammonia solution 3% analytical reagent, for determination of silicium dioxide
Supplier: Bernd Kraft
Certificates
About this item
CAS Number:1336-21-6
:7664-41-7
Formula:NH₃
MDL Number:MFCD00066650
:35.05 g/mol
EINECS:215-647-6
:GHS07
Storage Temperature:Ambient