Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Lead (II) nitrate
Lead(II) nitrate 0.1 mol/l (0.2 N) in aqueous solution Reag. Ph. Eur.
Lead(II) nitrate 0.1 mol/l (0.2 N) in aqueous solution Reag. Ph. Eur.
Lead(II) nitrate 0.1 mol/l (0.2 N) in aqueous solution Reag. Ph. Eur.
Supplier:
Bernd Kraft
Certificates
Expand image
Lead(II) nitrate 0.1 mol/l (0.2 N) in aqueous solution Reag. Ph. Eur.
Supplier:
Bernd Kraft
Certificates
About this item
CAS Number:
10099-74-8
:
Formula:
Pb(NO₃)₂
MDL Number:
MFCD00011153
:
331.21 g/mol
EINECS:
233-245-9
UN:
3082
ADR:
9,III
Related Information
Documentation
Certificates
Search by Lot Number
Documentation