n-Hexane for residue analysis
KRAF12650.4700
:
- Pack type:Glass bottle
- Pk:2.500 mL
- Seq:2895
:
:86.18 g/mol
:13,04712
:69 °C (1013 hPa)
:–94.3 °C
:0.659 g/cm³ (20 °C)
:–22 °C
:Ambient
:203-777-6
:1208
:3,II
:GHS02
:GHS08
:GHS07
:GHS09
:110-54-3
:92112-69-1
:H₃C(CH₂)₄CH₃
:MFCD00009520