Order Entry
Germany
ContactUsLinkComponent
Acetic acid ≥99% for trace analysis
Acetic acid ≥99% for trace analysis
Catalog # KRAF20693.2750
Supplier:  Bernd Kraft
CAS Number:  
undefined
Acetic acid ≥99% for trace analysis
Catalog # KRAF20693.2750
Supplier:  Bernd Kraft
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    0,5 L
  • Seq:
    5036

Specifications

About this item

 :
 :60.05 g/mol
Storage Temperature:Ambient
EINECS:200-580-7
UN:2789
ADR:8,II
 :GHS02
 :GHS05
CAS Number:64-19-7
Formula:H₃CCOOH
MDL Number:MFCD00036152