Order Entry
Germany
ContactUsLinkComponent
Zinc(II) nitrate hexahydrate ≥98%
Zinc(II) nitrate hexahydrate ≥98%
Catalog # ABCRAB122321.0100
Supplier:  abcr
CAS Number:  
Zinc(II) nitrate hexahydrate ≥98%
Catalog # ABCRAB122321.0100
Supplier:  abcr
CAS Number:  

Specifications

  • Pk:
    100 g

Specifications

About this item

 :
 :297.49 g/mol
Merck Index:12,10273
Boiling Pt:∼105 °C (1013 hPa)
Melting Pt:36.4 °C
Density:2.065
Storage Temperature:Ambient
EINECS:231-943-8
UN:1514
ADR:5.1,II
 :GHS03
 :GHS06
 :GHS07
CAS Number:10196-18-6
Formula:ZnN₂O₆·6H₂O
MDL Number:MFCD00149889