Tetrahydrofuran stabilised, Technical Grade
Supplier: Bernd Kraft
Certificates
About this item
CAS Number:109-99-9
:
Formula:C₄H₈O
MDL Number:MFCD00005356
:72.11 g/mol
Merck Index:12,09351
Boiling Pt:66 °C (1013 hPa)
Melting Pt:–108.5 °C
Density:0.888 g/cm³ (20 °C)
Flash Pt:–21.5 °C
UN:2056
ADR:3,II
:Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
Storage Temperature:Ambient
EINECS:203-726-8
REACH:05-2114615188-48
:GHS02
:GHS08
:GHS07