Order Entry
Germany
ContactUsLinkComponent
Perchloric acid 65 - 74% for trace analysis
Oxidizing
Corrosive
Perchloric acid 65 - 74% for trace analysis
Catalog # KRAF20696.2750
Supplier:  Bernd Kraft
CAS Number:  
undefined
Oxidizing
Corrosive
Perchloric acid 65 - 74% for trace analysis
Catalog # KRAF20696.2750
Supplier:  Bernd Kraft
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

About this item

 :
 :100.46 g/mol
Boiling Pt:160…181 °C (1013 hPa)
Melting Pt:–112.2 °C
Density:1.53…1.768 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-512-4
UN:1873
ADR:5.1,I
 :GHS03
 :GHS05
CAS Number:7601-90-3
Formula:HClO₄
MDL Number:MFCD00011325