Ethanol absolute, pure for molecular biology (200 Proof), Sigma-Aldrich®
SIALE7023-6X500ML
: Merck
:
- Pack type:Glass bottle
- Pk:3.000 mL
- Seq:2915
:
:46.07 g/mol
:12,03806
:78.3 °C (1013 hPa)
:–117 °C
:0.7895 g/cm³ (20 °C)
:12 °C
:200-578-6
:1170
:3,II
:M-Clarity™ quality level = MQ200
:01-2119457610-43
:GHS02
:Ambient
:64-17-5
:C₂H₅OH
:MFCD00003568