Ammonia solution 0.1 mol/l (0.1 N)
Supplier: Bernd Kraft
Certificates
About this item
CAS Number:1336-21-6
:7664-41-7
Formula:NH₃
MDL Number:MFCD00066650
:35.05 g/mol
Storage Temperature:Ambient
EINECS:215-647-6
Frequently Bought Together
Detergents, liquid, for automated cleaning of glassware, neodisher® FLA
Supplier: Dr. Weigert
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Neutralising agent for the automated cleaning of glassware, neodisher® Z
Supplier: Dr. Weigert
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
VWR® PH 20 pH Meter, Handheld
Supplier: VWR Collection
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
VWR®, Safety Pipette Fillers, Standard/Universal/Flip
Supplier: VWR Collection
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Ammonia solution 0.02 mol/l (0.02 N)
Supplier: Bernd Kraft