Order Entry
Germany
ContactUsLinkComponent
 
Boron standard solution, 1000 mg/L B in water (from B(OH₃)) standard for AAS
  KRAF03357.2000
 :  
Boron standard solution, 1000 mg/L B in water (from B(OH₃)) standard for AAS
  KRAF03357.2000
 :  

 

  • Concentration:
    1000 mg/L
  • Matrix:
    H₂O
  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Element:
    Boron
  • Symbol:
    B

 

 

 : Traceable to NIST