Beryllium standard solution, 1000 mg/L Be in dil. nitric acid (from Be) standard for AAS and ICP
Catalog # JOBIPLBE2-2T
Supplier: JOBIN YVON EMISSION
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:Nitric acid dil.
- Pack type:Glass bottle
- Pk:250 mL
- Element:Beryllium
- Symbol:Be
Specifications
About this item
Certifications: Traceable to SRM from NIST, tested in ISO 17034 / ISO 17025 accredited laboratory.