Order Entry
Germany
ContactUsLinkComponent
Arsenic standard solution, 1000 mg/L As in dil. nitric acid (from As) standard for AAS and ICP
Arsenic standard solution, 1000 mg/L As in dil. nitric acid (from As) standard for AAS and ICP
Catalog # JOBIPLAS2-2Y
CAS Number:  
Arsenic standard solution, 1000 mg/L As in dil. nitric acid (from As) standard for AAS and ICP
Catalog # JOBIPLAS2-2Y
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    2% HNO₃
  • Pack type:
    Glass bottle
  • Pk:
    125 mL
  • Element:
    Arsenic
  • Symbol:
    As

Specifications

About this item

Certifications: Traceable to SRM from NIST, tested in ISO 17034 / ISO 17025 accredited laboratory.