
Hexamethyldisiloxane, 0.65 cSt (25°C)
Catalog # ABCRAB111176.1500
Supplier: abcr
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1,5 kg
Specifications
About this item
:
:162.38 g/mol
Boiling Pt:101 °C (1013 hPa)
Melting Pt:–59 °C
Density:0.764 g/cm³ (20 °C)
Flash Pt:0.6 °C (closed cup)
Storage Temperature:Ambient
EINECS:203-492-7
UN:1993
ADR:3,II
REACH:01-2119496108-31
:GHS02
CAS Number:107-46-0
Formula:C₆H₁₈OSi₂
MDL Number:MFCD00008265