Order Entry
Germany
Orders LinkContactUsLinkComponent
Chloroform stabilised, Technical Grade

Chloroform stabilised, Technical Grade

Toxic
Health
Supplier:   Bernd Kraft
Chloroform stabilised, Technical Grade
Supplier:  Bernd Kraft
Certificates
Chloroform stabilised, Technical Grade
Chloroform stabilised, Technical Grade

2 Options Available Below

Product Details & Documents
Synonyms:  Methane, trichloro-, Trichloroform, Methylidyne trichloride, Trichloromethane
  • CAS Number:67-66-3
  • Formula:CHCl₃
  • MDL Number:MFCD00000826
  • Molecular Weight:119.38 g/mol
  • Merck Index:13,02160
  • Boiling Pt:61.7 °C (1013 hPa)
  • Melting Pt:–63 °C
  • Density:1.4832 g/cm³ (20 °C)
  • EINECS:200-663-8
  • ADR:6.1,III
  •  :Stabilised with ethanol
  • Storage Temperature:Ambient
  • UN:1888
  • REACH:05-2114614906-45
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    2.500 mL
    Log in for availability
    Log in for availability
    1 * 2.500 mL
    1 * 2.500 mL - 62,90 €
    62,90 €
    1 * 2.500 mL
       62,90 €

  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 37,50 €
    37,50 €
    1 * 1 L
       37,50 €