Octyltriethoxysilane
Catalog # ABCRAB110649.2000
Supplier: abcr
CAS Number:
Specifications
- Pk:2 kg
Specifications
About this item
:
:276.49 g/mol
Boiling Pt:265 °C (1013 hPa)
Melting Pt:–40 °C
Density:0.8515 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:220-941-2
:GHS07
CAS Number:2943-75-1
Formula:C₁₄H₃₂O₃Si
MDL Number:MFCD00039883