Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Acetic acid
Acetic acid 0.1 mol/l (0.1 N) Reag. Ph. Eur.
Acetic acid 0.1 mol/l (0.1 N) Reag. Ph. Eur.
Acetic acid 0.1 mol/l (0.1 N) Reag. Ph. Eur.
Supplier:
Bernd Kraft
Certificates
Expand image
Acetic acid 0.1 mol/l (0.1 N) Reag. Ph. Eur.
Supplier:
Bernd Kraft
About this item
CAS Number:
64-19-7
:
Formula:
H₃CCOOH
MDL Number:
MFCD00036152
:
60.05 g/mol
Storage Temperature:
Ambient
EINECS:
200-580-7