Order Entry
Germany
Orders LinkContactUsLinkComponent
Ammonia solution 32%, Technical Grade

Ammonia solution 32%, Technical Grade

Corrosive
Irritant
Environment
Supplier:   Bernd Kraft
Ammonia solution 32%, Technical Grade
Supplier:  Bernd Kraft
Certificates
Ammonia solution 32%, Technical Grade

3 Options Available Below

Product Details & Documents
Synonyms:  Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
  • CAS Number:1336-21-6
  • Formula:NH₃
  • MDL Number:MFCD00066650
  • Molecular Weight:35.05 g/mol
  • Boiling Pt:32…37.7 °C (1013 hPa)
  • Melting Pt:–60…–57.5 °C
  • Density:0.88…0.91 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-647-6
  • UN:2672
  • ADR:8,III
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 106,00 €
    106,00 €
    1 * 5 L
       106,00 €

  • Plastic bottle
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 188,00 €
    188,00 €
    1 * 10 L
       188,00 €

  • Plastic bottle
    25 L
    Log in for availability
    Log in for availability
    1 * 25 L
    1 * 25 L - 371,00 €
    371,00 €
    1 * 25 L
       371,00 €