Barium standard solution, 1,000 mg/L Ba in dil. nitric acid standard for ICP
Catalog # JOBIJYICP-BA
Supplier: JOBIN YVON EMISSION
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:Nitric acid dil.
- Pack type:Glass bottle
- Pk:100 mL
- Environmentally Preferable:
- Element:Barium
- Symbol:Ba