Order Entry
Germany
Orders LinkContactUsLinkComponent
Ammonia solution 20%, Technical Grade

Ammonia solution 20%, Technical Grade

Corrosive
Irritant
Environment
Supplier:   Bernd Kraft
Ammonia solution 20%, Technical Grade
Supplier:  Bernd Kraft
Certificates
Ammonia solution 20%, Technical Grade

3 Options Available Below

Product Details & Documents
Synonyms:  Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
  • CAS Number:1336-21-6
  • Formula:NH₃
  • MDL Number:MFCD00066650
  • Molecular Weight:35.05 g/mol
  • Density:0.92 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-647-6
  • UN:2672
  • ADR:8,III
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 74,10 €
    74,10 €
    1 * 5 L
       74,10 €

  • Plastic bottle
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 106,00 €
    106,00 €
    1 * 10 L
       106,00 €

  • Plastic bottle
    25 L
    Log in for availability
    Log in for availability
    1 * 25 L
    1 * 25 L - 185,00 €
    185,00 €
    1 * 25 L
       185,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.