Order Entry
Germany
ContactUsLinkComponent
Ammonium standard solution, 1000 mg/L NH₄ in water standard for Ion Chromatography
Ammonium standard solution, 1000 mg/L NH₄ in water standard for Ion Chromatography
Catalog # REGEICCS01
Supplier:  REAGECON
CAS Number:  
Ammonium standard solution, 1000 mg/L NH₄ in water standard for Ion Chromatography
Catalog # REGEICCS01
Supplier:  REAGECON
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    H₂O
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Description:
    Ammonium standard solution

Specifications

About this item