Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Acetic acid
Acetic acid 10%, extra pure
Acetic acid 10%, extra pure
Acetic acid 10%, extra pure
Supplier:
Bernd Kraft
Certificates
Expand image
Acetic acid 10%, extra pure
Supplier:
Bernd Kraft
About this item
CAS Number:
64-19-7
:
Formula:
H₃CCOOH
MDL Number:
MFCD00036152
:
60.05 g/mol
Storage Temperature:
Ambient
EINECS:
200-580-7
UN:
2790
ADR:
8,III
:
GHS07
Related Information
Documentation
Product specification
KRAF20152.4000_TDS_EN
Documentation