Order Entry
Germany
ContactUsLinkComponent
 
Cell spreaders, stainless steel
Catalog # HEATHS86655
Supplier:  HEATHROW
undefined
Cell spreaders, stainless steel
Catalog # HEATHS86655
Supplier:  HEATHROW

Specifications

  • Description:
    Cell spreader, stainless steel, blade width 33 mm
  • Length:
    203 mm
  • Pk:
    1

Specifications

About this item

Stainless steel spreader for use with 90 mm plates. Smooth triangular end makes spreading easy - the upturned end will not cut into agar.