Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Forceps
Tweezers, BOLA
Tweezers, BOLA
Tweezers, BOLA
Supplier:
Bohlender
Certificates
Expand image
Tweezers, BOLA
Supplier:
Bohlender
About this item
Precast tweezers made of PTFE.
With pointed or blunt ends
Universal chemical resistance
Temperature resistant from −200 to +250 °C