Screw caps, WHEATON®
About this item
Rubber lined caps feature a non-toxic white styrene-butadiene rubber liner ideal for cell culture work.
- Aluminum foil faced pulp board liners can be used with organic solvents. Good resistance to hydrocarbons, oils, ketones and alcohols Not good for acids or alkalis.
Use low density polyethylene (LDPE) lined caps with distilled water, analytical standards and reagents.
PTFE faced (14B) styrene-butadiene rubber liner provides a totally inert inner seal and surface facing the sample or product.
Bonded PTFE faced silicone liner ideal for storage of volatile solvents, sensitive compounds and corrosive chemicals.
Poly-Seal liner (LDPE) has unique cone liner design and offers superior torque retention and excellent sealing characteristics.
Caution: It is recommended that liner be tested prior to use for leak resistant applications.
62 Options Available Below
Product Details & Documents
Rubber lined caps feature a non-toxic white styrene-butadiene rubber liner ideal for cell culture work.
- Aluminum foil faced pulp board liners can be used with organic solvents. Good resistance to hydrocarbons, oils, ketones and alcohols Not good for acids or alkalis.
Use low density polyethylene (LDPE) lined caps with distilled water, analytical standards and reagents.
PTFE faced (14B) styrene-butadiene rubber liner provides a totally inert inner seal and surface facing the sample or product.
Bonded PTFE faced silicone liner ideal for storage of volatile solvents, sensitive compounds and corrosive chemicals.
Poly-Seal liner (LDPE) has unique cone liner design and offers superior torque retention and excellent sealing characteristics.
Caution: It is recommended that liner be tested prior to use for leak resistant applications.
Product Family Options
With 14B rubber liner
Product Information
- ThreadColourAvailabilityPrice
- 548-2150Screw cap, solid top, phenolic, with 14B rubber linerScrew cap, solid top, phenolic, with 14B rubber liner8425BlackLog in for availability1 * 1.000 ST364,00 €
- 548-2151Screw cap, solid top, phenolic, with 14B rubber linerScrew cap, solid top, phenolic, with 14B rubber liner13425BlackLog in for availability1 * 200 ST114,00 €
- 548-2152Screw cap, solid top, phenolic, with 14B rubber linerScrew cap, solid top, phenolic, with 14B rubber liner15425BlackLog in for availability1 * 200 ST130,00 €
- 548-2153Screw cap, solid top, phenolic, with 14B rubber linerScrew cap, solid top, phenolic, with 14B rubber liner18400BlackLog in for availability1 * 500 ST369,00 €
- WHEA240264Screw cap, solid top, phenolic, with 14B rubber linerScrew cap, solid top, phenolic, with 14B rubber liner18415BlackLog in for availability1 * 500 ST323,00 €
- WHEAW239303Screw cap, solid top, phenolic, with 14B rubber linerScrew cap, solid top, phenolic, with 14B rubber liner38-400BlackLog in for availability1 * 72 ST170,00 €
- 548-2154Screw cap, solid top, phenolic, with 14B rubber linerScrew cap, solid top, phenolic, with 14B rubber liner20400BlackLog in for availability1 * 500 ST599,00 €
- WHEAW239298Screw cap, solid top, phenolic, with 14B rubber linerScrew cap, solid top, phenolic, with 14B rubber liner20-400BlackLog in for availability1 * 144 ST130,00 €
- WHEAW239299Screw cap, solid top, phenolic, with 14B rubber linerScrew cap, solid top, phenolic, with 14B rubber liner22400BlackLog in for availability1 * 144 ST149,00 €
- 548-2155Screw cap, solid top, phenolic, with 14B rubber linerScrew cap, solid top, phenolic, with 14B rubber liner24400BlackLog in for availability1 * 500 ST705,00 €
- WHEAW239300Screw cap, solid top, phenolic, with 14B rubber linerScrew cap, solid top, phenolic, with 14B rubber liner24400BlackLog in for availability1 * 144 ST163,00 €
- WHEAW239301Screw cap, solid top, phenolic, with 14B rubber linerScrew cap, solid top, phenolic, with 14B rubber liner28-400BlackLog in for availability1 * 144 ST179,00 €
- WHEA240280Screw cap, solid top, phenolic, with 14B rubber linerScrew cap, solid top, phenolic, with 14B rubber liner33430BlackLog in for availability1 * 200 ST451,00 €
- WHEAW239302Screw cap, solid top, phenolic, with 14B rubber linerScrew cap, solid top, phenolic, with 14B rubber liner33-400BlackLog in for availability1 * 144 ST222,00 €
- WHEA240223Screw cap, solid top, phenolic, with 14B rubber linerScrew cap, solid top, phenolic, with 14B rubber liner38400BlackLog in for availability1 * 200 ST425,00 €
- WHEA240269Screw cap, solid top, phenolic, with 14B rubber linerScrew cap, solid top, phenolic, with 14B rubber liner38415BlackLog in for availability1 * 200 ST995,00 €
- WHEA240281Screw cap, solid top, phenolic, with 14B rubber linerScrew cap, solid top, phenolic, with 14B rubber liner38430BlackLog in for availability1 * 200 ST573,00 €
- WHEAW239304Screw cap, solid top, phenolic, with 14B rubber linerScrew cap, solid top, phenolic, with 14B rubber liner43-400BlackLog in for availability1 * 72 ST181,00 €
- WHEA240228Screw cap, solid top, phenolic, with 14B rubber linerScrew cap, solid top, phenolic, with 14B rubber liner51400BlackLog in for availability1 * 100 ST318,00 €
With PTFE faced 14B styrene-butadiene rubber liner
Product Information
- ThreadColourAvailabilityPrice
- WHEAW240406Screw cap, solid top, phenolic, with PTFE faced 14B styrene-butadiene rubber linerScrew cap, solid top, phenolic, with PTFE faced 14B styrene-butadiene rubber liner8-425BlackLog in for availability1 * 200 ST327,00 €
- 548-2030Screw cap, solid top, phenolic, with PTFE faced 14B styrene-butadiene rubber linerScrew cap, solid top, phenolic, with PTFE faced 14B styrene-butadiene rubber liner13-425BlackLog in for availability1 * 200 ST337,00 €
- WHEAW240820Screw cap, solid top, phenolic, with PTFE faced 14B styrene-butadiene rubber linerScrew cap, solid top, phenolic, with PTFE faced 14B styrene-butadiene rubber liner13-425BlackLog in for availability1 * 250 ST388,00 €
- 548-2031Screw cap, solid top, phenolic, with PTFE faced 14B styrene-butadiene rubber linerScrew cap, solid top, phenolic, with PTFE faced 14B styrene-butadiene rubber liner15-425BlackLog in for availability1 * 200 ST365,00 €
- WHEAW240822Screw cap, solid top, phenolic, with PTFE faced 14B styrene-butadiene rubber linerScrew cap, solid top, phenolic, with PTFE faced 14B styrene-butadiene rubber liner15-425BlackLog in for availability1 * 250 ST425,00 €
- WHEA240463Screw cap, solid top, phenolic, with PTFE faced 14B styrene-butadiene rubber linerScrew cap, solid top, phenolic, with PTFE faced 14B styrene-butadiene rubber liner15-415BlackLog in for availability1 * 200 ST384,00 €
- WHEA240414Screw cap, solid top, phenolic, with PTFE faced 14B styrene-butadiene rubber linerScrew cap, solid top, phenolic, with PTFE faced 14B styrene-butadiene rubber liner18-415BlackLog in for availability1 * 200 ST417,00 €
- 548-2032Screw cap, solid top, phenolic, with PTFE faced 14B styrene-butadiene rubber linerScrew cap, solid top, phenolic, with PTFE faced 14B styrene-butadiene rubber liner18-400BlackLog in for availability1 * 200 ST506,00 €
- 548-2033Screw cap, solid top, phenolic, with PTFE faced 14B styrene-butadiene rubber linerScrew cap, solid top, phenolic, with PTFE faced 14B styrene-butadiene rubber liner20-400BlackLog in for availability1 * 100 ST346,00 €
- WHEAW240824Screw cap, solid top, phenolic, with PTFE faced 14B styrene-butadiene rubber linerScrew cap, solid top, phenolic, with PTFE faced 14B styrene-butadiene rubber liner22-400BlackLog in for availability1 * 100 ST337,00 €
- WHEAW240825Screw cap, solid top, phenolic, with PTFE faced 14B styrene-butadiene rubber linerScrew cap, solid top, phenolic, with PTFE faced 14B styrene-butadiene rubber liner22-400BlackLog in for availability1 * 500 ST1 450,00 €
- 548-2034Screw cap, solid top, phenolic, with PTFE faced 14B styrene-butadiene rubber linerScrew cap, solid top, phenolic, with PTFE faced 14B styrene-butadiene rubber liner24-400BlackLog in for availability1 * 100 ST435,00 €
- WHEA240419Screw cap, solid top, phenolic, with PTFE faced 14B styrene-butadiene rubber linerScrew cap, solid top, phenolic, with PTFE faced 14B styrene-butadiene rubber liner28-400BlackLog in for availability1 * 100 ST423,00 €
- 548-2035Screw cap, solid top, phenolic, with PTFE faced 14B styrene-butadiene rubber linerScrew cap, solid top, phenolic, with PTFE faced 14B styrene-butadiene rubber liner33-430BlackLog in for availability1 * 100 ST574,00 €
- WHEA240421Screw cap, solid top, phenolic, with PTFE faced 14B styrene-butadiene rubber linerScrew cap, solid top, phenolic, with PTFE faced 14B styrene-butadiene rubber liner33-400BlackLog in for availability1 * 100 ST546,00 €
- WHEA240481Screw cap, solid top, phenolic, with PTFE faced 14B styrene-butadiene rubber linerScrew cap, solid top, phenolic, with PTFE faced 14B styrene-butadiene rubber liner38-430BlackLog in for availability1 * 100 ST631,00 €
- WHEAW240823Screw cap, solid top, phenolic, with PTFE faced 14B styrene-butadiene rubber linerScrew cap, solid top, phenolic, with PTFE faced 14B styrene-butadiene rubber liner15-425BlackLog in for availability1 * 1.000 ST1 510,00 €
With PE Poly-Seal® liner
Product Information
- ThreadColourAvailabilityPrice
- WHEA239249Screw cap, solid top, phenolic, with PE Poly-Seal® linerScrew cap, solid top, phenolic, with PE Poly-Seal® liner13-425BlackLog in for availability1 * 144 ST98,10 €
- WHEA239250Screw cap, solid top, phenolic, with PE Poly-Seal® linerScrew cap, solid top, phenolic, with PE Poly-Seal® liner15-425BlackLog in for availability1 * 144 ST106,00 €
- WHEA239251Screw cap, solid top, phenolic, with PE Poly-Seal® linerScrew cap, solid top, phenolic, with PE Poly-Seal® liner18-400BlackLog in for availability1 * 144 ST109,00 €
- WHEA239451Screw cap, solid top, phenolic, with PE Poly-Seal® linerScrew cap, solid top, phenolic, with PE Poly-Seal® liner18-400BlackLog in for availability1 * 6.500 ST3 820,00 €
- WHEA239253Screw cap, solid top, phenolic, with PE Poly-Seal® linerScrew cap, solid top, phenolic, with PE Poly-Seal® liner20-400BlackLog in for availability1 * 144 ST101,00 €
- WHEA239255Screw cap, solid top, phenolic, with PE Poly-Seal® linerScrew cap, solid top, phenolic, with PE Poly-Seal® liner22-400BlackLog in for availability1 * 144 ST110,00 €
- WHEA239257Screw cap, solid top, phenolic, with PE Poly-Seal® linerScrew cap, solid top, phenolic, with PE Poly-Seal® liner24-400BlackLog in for availability1 * 144 ST113,00 €
- WHEA239259Screw cap, solid top, phenolic, with PE Poly-Seal® linerScrew cap, solid top, phenolic, with PE Poly-Seal® liner28-400BlackLog in for availability1 * 144 ST130,00 €
- WHEA240119Screw cap, solid top, phenolic, with PE Poly-Seal® linerScrew cap, solid top, phenolic, with PE Poly-Seal® liner28-400BlackLog in for availability1 * 100 ST188,00 €
- WHEA239260Screw cap, solid top, phenolic, with PE Poly-Seal® linerScrew cap, solid top, phenolic, with PE Poly-Seal® liner33-400BlackLog in for availability1 * 144 ST165,00 €
- WHEA240121Screw cap, solid top, phenolic, with PE Poly-Seal® linerScrew cap, solid top, phenolic, with PE Poly-Seal® liner33-400BlackLog in for availability1 * 100 ST217,00 €
- WHEA240181Screw cap, solid top, phenolic, with PE Poly-Seal® linerScrew cap, solid top, phenolic, with PE Poly-Seal® liner38-430BlackLog in for availability1 * 100 ST253,00 €
With aluminum foil faced pulp board liner
Product Information
- ThreadColourAvailabilityPrice
- WHEA240319Screw cap, solid top, phenolic, with aluminum foil faced pulp board linerScrew cap, solid top, phenolic, with aluminum foil faced pulp board liner28-400BlackLog in for availability1 * 100 ST165,00 €
- WHEA240321Screw cap, solid top, phenolic, with aluminum foil faced pulp board linerScrew cap, solid top, phenolic, with aluminum foil faced pulp board liner33-400BlackLog in for availability1 * 100 ST169,00 €
- WHEA240381Screw cap, solid top, phenolic, with aluminum foil faced pulp board linerScrew cap, solid top, phenolic, with aluminum foil faced pulp board liner38-430BlackLog in for availability1 * 200 ST547,00 €
- WHEA240330Screw cap, solid top, phenolic, with aluminum foil faced pulp board linerScrew cap, solid top, phenolic, with aluminum foil faced pulp board liner58-400BlackLog in for availability1 * 200 ST608,00 €
With PE (LDPE) liner
Product Information
- ThreadColourAvailabilityPrice
- WHEA240080Screw cap, solid top, phenolic, with PE (LDPE) linerScrew cap, solid top, phenolic, with PE (LDPE) liner33-430BlackLog in for availability1 * 200 ST445,00 €
- WHEA240081Screw cap, solid top, phenolic, with PE (LDPE) linerScrew cap, solid top, phenolic, with PE (LDPE) liner38-430BlackLog in for availability1 * 200 ST513,00 €
With gray chlorobutyl/50 septa and flange
Product Information
- ThreadColourAvailabilityPrice
- WHEA240680Screw cap, open top, phenolic, with gray chlorobutyl/50 septa and flangeScrew cap, open top, phenolic, with gray chlorobutyl/50 septa and flange33-430BlackLog in for availability1 * 100 ST304,00 €
Without liner
Product Information
- ThreadColourAvailabilityPrice
- WHEAW240508Screw cap, open top, phenolic, without liner
- WHEAW240509Screw cap, open top, phenolic, without liner
- WHEAW240516Screw cap, open top, phenolic, without liner
- WHEAW240517Screw cap, open top, phenolic, without liner
- WHEA239853Screw cap, solid top, phenolic, without liner
- WHEAW240518Screw cap, open top, phenolic, without liner
- 218-2147Screw cap, open top, phenolic, without liner