Order Entry
Germany
ContactUsLinkComponent
 
illustra™ GFX™ 96 Purification kit
  28-9034-45
 :  Cytiva
 :  
undefined
illustra™ GFX™ 96 Purification kit
  28-9034-45
 :  Cytiva
 :  

 

  • Pk:
    1 KIT
  • Method / Format:
    Multiwell plate
  • Prep size:
    Mini
  • Refine target molecule:
    Amplified/modified DNA
  • Sample type:
    PCR and other enzymatic reactions
  • Target molecule:
    DNA
  • Description:
    illustra™ PCR GFX™ 96 Purification Kit, 10×96-well plates
  • Description:
    illustra™ PCR GFX™ 96 Purification Kit

 

 

The illustra™ GFX™ 96 Purification Kit utilises glass fibre matrix technology in a 96-well format for highly efficient purification of PCR products. Fragments from PCR are captured by the matrix in the presence of a chaotropic salt, and contaminants are conveniently removed by washing the matrix with a buffered ethanol solution.

  • For the purification of up to 96 PCR products (0,1 to 10 kb) simultaneously in as little as 15 minutes
  • High yields of pure DNA recovered in a small volume of water or a low-ionic strength buffer
  • Typical recoveries are >85% for PCR products 100 bp to 10 kb in length; salt removal typically >99%
  • Avoids ethanol precipitations and hazardous organic extractions

Purified DNA is ready for use in most applications, including fluorescent sequencing, microarrays, labelling, hybridisation, ligation, and transformation.

 : Kit contains sufficient quantities of binding plates (10), wash plates (10), collection plates (10), capture buffer (blue, 288 ml), wash buffer (yellow, 80 ml), a choice of two elution buffers each 48 ml (Tris-HCl and sterile water) and an instruction booklet.