Orders LinkContactUsLinkComponent
dNTP solutions, illustra™
dNTP solutions, illustra™
Supplier:   Cytiva
dNTP solutions, illustra™
Supplier:  Cytiva
Certificates
dNTP solutions, illustra™
dNTP solutions, illustra™
7 Options Available Below
Product Details & Documents
illustra™ dNTPs are high purity solutions for enzymatically sensitive applications, such as nucleic acid amplification, sequencing, labelling, mutagenesis, cDNA synthesis and expression profiling.
  • Free from DNase, RNase, and nicking enzyme activity
  • Greater than 99% triphosphate purity (by HPLC) for optimum performance and consistency
  • Buffer free and ready-to-use solutions in multiple formats
  • Functionally tested for long PCR and sequencing
Product Family Options
Product Information
  • Pk
    Availability
    Price
  • 28-4065-42
    dUTP, 100 mM solution
    dUTP, 100 mM solution
    100 µmol
    Log in for availability
    Log in for availability
    1 * 100 µmol
    1 * 100 µmol - 170,00 €
    170,00 €
    1 * 100 µmol
       170,00 €
  • 28-4065-41
    dUTP, 100 mM solution
    dUTP, 100 mM solution
    25 µmol
    Log in for availability
    Log in for availability
    1 * 25 µmol
    1 * 25 µmol - 53,50 €
    53,50 €
    1 * 25 µmol
       53,50 €
  • 28-4065-32
    dTTP, 100 mMol solution
    dTTP, 100 mMol solution
    100 µmol
    Log in for availability
    Log in for availability
    1 * 100 µmol
    1 * 100 µmol - 170,00 €
    170,00 €
    1 * 100 µmol
       170,00 €
  • 28-4065-22
    dGTP, 100 mMol solution
    dGTP, 100 mMol solution
    100 µmol
    Log in for availability
    Log in for availability
    1 * 100 µmol
    1 * 100 µmol - 170,00 €
    170,00 €
    1 * 100 µmol
       170,00 €
  • 28-4065-02
    dATP, 100 mMol solution
    dATP, 100 mMol solution
    100 µmol
    Log in for availability
    Log in for availability
    1 * 100 µmol
    1 * 100 µmol - 170,00 €
    170,00 €
    1 * 100 µmol
       170,00 €
  • 28-4065-01
    dATP, 100 mMol solution
    dATP, 100 mMol solution
    25 µmol
    Log in for availability
    Log in for availability
    1 * 25 µmol
    1 * 25 µmol - 53,50 €
    53,50 €
    1 * 25 µmol
       53,50 €
  • 28-4065-12
    dCTP, 100 mMol solution
    dCTP, 100 mMol solution
    100 µmol
    Log in for availability
    Log in for availability
    1 * 100 µmol
    1 * 100 µmol - 170,00 €
    170,00 €
    1 * 100 µmol
       170,00 €