Order Entry
Germany
ContactUsLinkComponent
illustra™ ExoProStar™ 1-STEP kits
illustra™ ExoProStar™ 1-STEP kits
Supplier:  Cytiva
Certificates
undefined
illustra™ ExoProStar™ 1-STEP kits
Supplier:  Cytiva
Certificates

Specifications

  • Method / Format:
    Enzymatic clean-up
  • Prep size:
    Scalable
  • Refine target molecule:
    Amplified/modified DNA
  • Sample type:
    PCR and other enzymatic reactions
  • Target molecule:
    DNA
  • Description:
    illustra™ ExoProStar™ 1-STEP Kit

Specifications

Related Information