ContactUsLinkComponent
Stirrers, Corning® Gosselin™
Stirrers, Corning® Gosselin™
Catalog # GOSSAGI-06
Supplier:  Corning
   Low Manufacturing Impact
undefined
Stirrers, Corning® Gosselin™
Catalog # GOSSAGI-06
Supplier:  Corning
   Low Manufacturing Impact
Specifications
  • Description:
    Stirrer, sterile SAL 10-3
  • Length:
    150 mm
  • Material:
    PS
  • Seq:
    02
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    RECs, ISO 14001 - Borre
  • Pk:
    5.000
Specifications
About this item
These PS stirrer are ideal for use in the dairy, environmental, microbiology and industry sectors. Colour: white.