General purpose gloves, Tricoton Ultralight
Supplier: HONEYWELL SAFETY
Certificates
About this item
These lightweight cotton knitted gloves limits the penetration of dirt while offering protection against small cuts.
- Knitting technique helps to obtain a glove that conforms snugly to the hand and good dexterity
- Maximum comfort and added sturdiness
- Pure cotton has a neutral pH level: skin-friendly, limits the absorption of perspiration
- Light colour enhances the glove's hygiene and cleanliness properties
Ideal for fine handlings in clean environments. Can also be used as undergloves.
Certifications: Cat. I.
Specifications
- Catalog No:
- DALLRGT350/9
- DALLRGT350/6
- DALLRGT350/8
- DALLRGT350/7
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Gauge:
- 13
- 13
- 13
- 13
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Cuff length:
- 6 cm
- 6 cm
- 6 cm
- 6 cm