Jumbo syringes
Supplier: Trajan Scientific and Medical
Certificates
About this item
These syringes are ideal for calibration of medical equipment such as respirators and spirometers, and for stack and air sampling.
- Designed for holding and dispensing large volumes of gas
- Heavy duty acrylic barrels
- Easy access to sample for the addition of standards or removal of subsample via secondary port
- Plunger stem can be unscrewed for ease of transportation and storage